R&D Systems
Manufacturer Code:MAB38951SP
Catalog # MAB38951SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 387310 |
Immunogen | NS0 mouse myeloma cell line transfected with human KOR Met1-Val380 Accession # P41145 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: K-OR-1; kappa opioid receptor; kappa opioid receptor kappa-type opioid receptor KOR K-OR-1 KOR-1 Opiate receptor kappa-1 opioid receptor kappa 1 OPRK OPRK1 Oprk2 R21; Kappa-type opioid receptor; Opiate receptor, kappa-1; opioid receptor, kappa 1
Gene Aliases: K-OR-1; KOR; KOR-1; OPRK; OPRK1
UniProt ID: (Human) P41145
Entrez Gene ID: (Human) 4986
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.