Novus Biologicals
Manufacturer Code:NB11079876
Catalog # NB11079876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K-OR-1; kappa opioid receptor; kappa opioid receptor kappa-type opioid receptor KOR K-OR-1 KOR-1 Opiate receptor kappa-1 opioid receptor kappa 1 OPRK; Kappa-type opioid receptor; Opiate receptor, kappa-1; opioid receptor, kappa 1
Gene Aliases: K-OR-1; KOR; KOR-1; OPRK; OPRK1
UniProt ID: (Human) P41145
Entrez Gene ID: (Human) 4986
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.