Invitrogen
Manufacturer Code:PA557490
Catalog # PIPA557490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KMT2D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALL1-related protein; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; Kabuki make-up syndrome; Kabuki mental retardation syndrome; lysine (K)-specific methyltransferase 2D; Lysine (K)-Specific Methyltransferase 2D ALR MLL2 MLL4; Lysine N-methyltransferase 2D; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; trinucleotide repeat containing 21
Gene Aliases: AAD10; ALR; CAGL114; KABUK1; KMS; KMT2D; MLL2; MLL4; TNRC21
UniProt ID: (Human) O14686
Entrez Gene ID: (Human) 8085
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.