Novus Biologicals
Manufacturer Code:NB600256G
Catalog # NB600256G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 2725 and 2775 of human myeloid/lymphoid or mixed-lineage leukemia 1 using the number given in Swiss-Prot entry Q03164 (GeneID 4297). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALL-1; ALL1 ALL-1MLL/GAS7 CXXC7TET1-MLL CXXC-type zinc finger protein 7 EC 2.1.1.43 HRXFLJ11783 HTRX HTRX1MLL-AF4 der(11) fusion protein KMT2ACDK6/MLL fusion protein Lysine N-methyltransferase 2A MLL/GAS7 fusion protein MLL/GMPS fusion protein MLL1 MLL1A myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog) myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog Drosophila) Trithorax-like protein TRX1histone-lysine N-methyltransferase MLL Zinc finger protein HRX; C-terminal cleavage product of 180 kDa; CDK6/MLL fusion protein; CXXC-type zinc finger protein 7; Histone-lysine N-methyltransferase 2A; lysine (K)-specific methyltransferase 2A; Lysine N-methyltransferase 2A; mixed lineage leukemia 1; MLL cleavage product C180; MLL cleavage product N320; MLL-AF4 der(11) fusion protein; MLL/ENL fusion protein; MLL/GAS7 fusion protein; MLL/GMPS fusion protein; MLL/hCDCrel fusion protein; Myeloid/lymphoid or mixed-lineage leukemia; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); Myeloid/lymphoid or mixed-lineage leukemia protein 1; N-terminal cleavage product of 320 kDa; p180; p320; rearranged MLL protein; Trithorax-like protein; Zinc finger protein HRX
Gene Aliases: ALL-1; ALL1; CXXC7; HRX; HTRX; HTRX1; KMT2A; MLL; MLL-AF9; MLL/GAS7; MLL1; MLL1A; TET1-MLL; TRX1; WDSTS
UniProt ID: (Human) Q03164
Entrez Gene ID: (Human) 4297
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.