Novus Biologicals
Manufacturer Code:NBP298967AF532
Catalog # NB056890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human KMT1A/SUV39H1. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H3-K9-HMTase 1; H3-K9-HMTase 1 Histone H3-K9 methyltransferase 1 histone-lysine N-methyltransferase SUV39H1 histone-lysine N-methyltransferase H3 lysine-9 specific 1 KMT1AEC 2.1.1.43 Lysine N-methyltransferase 1A MG44 Position-effect variegation 3-9 homolog Su(var)3-9 homolog 1 suppressor of variegation 3-9 (Drosophila) homolog 1 Suppressor of variegation 3-9 homolog 1 suppressor of variegation 3-9 homolog 1 (Drosophila) Suppressor of variegation 3-9 homolog 1 Suvar) 3-9 homolog SUV39H; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; Lysine N-methyltransferase 1A; Position-effect variegation 3-9 homolog; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1
Gene Aliases: H3-K9-HMTase 1; KMT1A; MG44; SUV39H; SUV39H1
UniProt ID: (Human) O43463
Entrez Gene ID: (Human) 6839
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.