Proteintech
Manufacturer Code:215101AP150UL
Catalog # 50-173-3456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLRF1 Fusion Protein Ag16008 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating coreceptor NKp80; Activating coreceptor NKp80 CLEC5C KLRF1 Lectin like receptor F1 ML; C-type lectin domain family 5 member C; killer cell lectin-like receptor F1; Killer cell lectin-like receptor subfamily F member 1; killer cell lectin-like receptor subfamily F, member 1; Lectin-like receptor F1
Gene Aliases: CLEC5C; KLRF1; ML; NKp80
UniProt ID: (Human) Q9NZS2
Entrez Gene ID: (Human) 51348
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.