Proteintech
Manufacturer Code:279911AP150UL
Catalog # 50-173-5802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLKB1 Fusion Protein Ag27491 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen, prostate specific; Fletcher factor; Fletcher factor Kininogenin KLK3 KLKB1 Plasma kallikrein Plasma prekallikrein PPK; kallikrein 3, plasma; kallikrein B, plasma (Fletcher factor) 1; Kininogenin; PKK; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein
Gene Aliases: APS; Kal-3; Kal3; Kalp1; KALP15; KLK3; KLKB1; Pk; PKK; PKKD; PPK; PSA
UniProt ID: (Human) P03952, (Mouse) P26262
Entrez Gene ID: (Human) 3818, (Rat) 25048, (Mouse) 16621
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.