OriGene
Manufacturer Code:TA506135
Catalog # 50-167-6483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10D3 |
Immunogen | Full length human recombit protein of human KLK8 produced in HEk293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hK8; HNP NP NRPN PRSS19 TADG14; Kallikrein-8; kallikrein-related peptidase 8; Neuropsin; NP; Ovasin; Serine protease 19; Serine protease TADG-14; Tumor-associated differentially expressed gene 14 protein
Gene Aliases: HNP; KLK8; NP; NRPN; PRSS19; TADG14; UNQ283/PRO322
UniProt ID: (Human) O60259
Entrez Gene ID: (Human) 11202
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.