Invitrogen
Manufacturer Code:PA544515
Catalog # PIPA544515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KLHL7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain containing protein; kelch-like 6; kelch-like 7; kelch-like family member 7; Kelch-like protein 7; kelch/BTB
Gene Aliases: 2700038B03Rik; D5Ertd363e; KLHL6; KLHL7; SBBI26
UniProt ID: (Human) Q8IXQ5, (Mouse) Q8BUL5, (Rat) Q5XHZ6
Entrez Gene ID: (Human) 55975, (Bovine) 534697, (Rabbit) 100338490, (Dog) 475255, (Horse) 100053660, (Guinea pig) 100720424, (Mouse) 52323, (Rat) 362303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.