Novus Biologicals
Manufacturer Code:NBP17059420UL
Catalog # NBP17059420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to KLHL31(kelch-like 31 (Drosophila)) The peptide sequence was selected from the C terminal of KLHL31. Peptide sequence TPRGWHCAVTLSDRVYVMGGSQLGPRGERVDVLTVECYSPATGQWSYAAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA345L23.2 BKLHD6 BTB and kelch domain-containing protein 6 KBTBD1 kelch repeat and BTB (POZ) domain containing 1 kelch repeat and BTB domain-containing protein 1 kelch-like 31 (Drosophila) kelch-like protein 31 kelch-like protein KLHL KLHL; BTB and kelch domain-containing protein 6; kelch repeat and BTB (POZ) domain containing 1; Kelch repeat and BTB domain-containing protein 1; kelch-like family member 31; Kelch-like protein 31; Kelch-like protein KLHL
Gene Aliases: bA345L23.2; BKLHD6; KBTBD1; KLHL; KLHL31
UniProt ID: (Human) Q9H511
Entrez Gene ID: (Human) 401265
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.