Proteintech
Manufacturer Code:169511AP150UL
Catalog # 50-173-1288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLHL3 Fusion Protein Ag10580 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kelch like 3 (Drosophila) Kelch like protein 3 KIAA1129 KLHL3; kelch-like 3; kelch-like family member 3; Kelch-like protein 3
Gene Aliases: 7530408C15Rik; AI430941; EG627648; KIAA1129; KLHL3; PHA2D; RGD1565218
UniProt ID: (Human) Q9UH77
Entrez Gene ID: (Human) 26249, (Mouse) 100503085, (Rat) 498697
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.