Invitrogen
Manufacturer Code:PA525162
Catalog # PIPA525162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 182-211 amino acids from the central region of human KLHL24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRE1 KRIP6 kainate receptor interacting protein for GluR6 kainate receptor-interacting protein for GluR6 kelch-like protein 24; kainate receptor interacting protein for GluR6; Kainate receptor-interacting protein for GluR6; kelch-like 24; kelch-like family member 24; Kelch-like protein 24; KRIP6; Protein DRE1
Gene Aliases: DRE1; KLHL24; KRIP6
UniProt ID: (Human) Q6TFL4
Entrez Gene ID: (Human) 54800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.