Novus Biologicals
Manufacturer Code:NBP271695C
Catalog # NBP271695C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G7 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ABP-KELCH Actin-binding protein Mayven kelch kelch (Drosophila)-like 2 (Mayven) kelch-like 2 Mayven (Drosophila) kelch-like protein 2 MAV MAYVEN; Actin-binding protein Mayven; kelch; kelch-like 2, Mayven; kelch-like family member 2; Kelch-like protein 2
Gene Aliases: ABP-KELCH; KLHL2; MAV; MAYVEN
UniProt ID: (Human) O95198
Entrez Gene ID: (Human) 11275
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.