Proteintech
Manufacturer Code:144491AP150UL
Catalog # 50-172-9459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLHL2 Fusion Protein Ag5950 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABP KELCH Actin binding protein Mayven Kelch like protein 2 KLHL2 MAV MAYVEN; Actin-binding protein Mayven; kelch; kelch-like 2, Mayven; kelch-like family member 2; Kelch-like protein 2; klhl2; mayven
Gene Aliases: 6030411N21Rik; ABP-KELCH; AU020744; KLHL2; MAV; MAYVEN
UniProt ID: (Human) O95198, (Mouse) Q8JZP3
Entrez Gene ID: (Human) 11275, (Mouse) 77113, (Rat) 290692
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.