Proteintech
Manufacturer Code:148491AP150UL
Catalog # 50-172-9775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kelch like 14 (Drosophila) Kelch like protein 14 KIAA1384 KLHL14; kelch-like 14; kelch-like family member 14; Kelch-like protein 14; Printor; Protein interactor of Torsin-1A; protein interactor of torsinA
Gene Aliases: 6330403N15Rik; AI785250; KIAA1384; KLHL14; printor; RGD1566178
UniProt ID: (Human) Q9P2G3, (Mouse) Q69ZK5
Entrez Gene ID: (Human) 57565, (Rat) 364823, (Mouse) 225266
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.