Novus Biologicals
Manufacturer Code:NBP217079
Catalog # NBP217079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human KLHL13. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB and kelch domain containing 2; BTB and kelch domain containing 2 BTB and kelch domain-containing protein 2 kelch-like 13 (Drosophila) kelch-like protein 13 KIAA1309 MGC74791; BTB and kelch domain-containing protein 2; kelch-like 13; kelch-like family member 13; Kelch-like protein 13
Gene Aliases: BKLHD2; KIAA1309; KLHL13
UniProt ID: (Human) Q9P2N7
Entrez Gene ID: (Human) 90293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.