Novus Biologicals
Manufacturer Code:NBP245511
Catalog # NBP245511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8A11 |
Immunogen | Full length human recombinant protein of human KLF9(NP_001197) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: basic transcription element binding protein 1; basic transcription element binding protein 1 Basic transcription element-binding protein 1 BTEB1GC-box-binding protein 1 BTEBBTE-binding protein 1 Krueppel-like factor 9 Kruppel-like factor 9 Transcription factor BTEB1; Basic transcription element-binding protein 1; BTE-binding protein 1; GC-box-binding protein 1; Krueppel-like factor 9; Kruppel-like factor 9; Transcription factor BTEB1
Gene Aliases: BTEB; BTEB1; KLF9
UniProt ID: (Human) Q16196
Entrez Gene ID: (Human) 687
Molecular Function:
DNA binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.