Novus Biologicals
Manufacturer Code:NBP266918
Catalog # NBP266918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF98-08 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: endothelial Kruppel-like zinc finger protein; Epithelial zinc finger protein EZF; Epithelial zinc finger protein EZF EZFendothelial Kruppel-like zinc finger protein GKLFKrueppel-like factor 4 Gut-enriched krueppel-like factor Kruppel-like factor 4 (gut); gut Kruppel-like factor; Gut-enriched krueppel-like factor; Krueppel-like factor 4; Kruppel-like factor 4 (gut)
Gene Aliases: EZF; GKLF; KLF4
UniProt ID: (Human) O43474
Entrez Gene ID: (Human) 9314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.