Abnova
Manufacturer Code:MAB10715
Catalog # 89-108-918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E5 |
Immunogen | Recombinant protein corresponding to human KLF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant KLF4.
For Research Use Only
Protein Aliases: endothelial Kruppel-like zinc finger protein; Epithelial zinc finger protein EZF; EZF; GKLF; gut Kruppel-like factor; Gut-enriched krueppel-like factor; Krueppel-like factor 4; Kruppel-like factor 4 (gut)
Gene Aliases: EZF; GKLF; KLF4
UniProt ID: (Human) O43474
Entrez Gene ID: (Human) 9314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.