OriGene
Manufacturer Code:TA807006
Catalog # 50-168-1353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F1 |
Immunogen | Full length human recombit protein of human KLF2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Krueppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; LKLF; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor
Gene Aliases: KLF2; LKLF
UniProt ID: (Human) Q9Y5W3
Entrez Gene ID: (Human) 10365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.