R&D Systems
Manufacturer Code:MAB5466
Catalog # MAB5466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 665333 |
Immunogen | <em>E. coli</em>-derived recombinant human KLF2 Pro71-Pro168 (predicted) Accession # Q9Y5W3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Krueppel-like factor 2; Krueppel-like factor 2 Kruppel-like factor 2 (lung) Kruppel-like factor LKLF LKLF LKLFLung krueppel-like factor lung Kruppel-like zinc finger transcription factor; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor
Gene Aliases: KLF2; LKLF
UniProt ID: (Human) Q9Y5W3
Entrez Gene ID: (Human) 10365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.