Invitrogen
Manufacturer Code:PA572410
Catalog # PIPA572410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 251-355/355 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Krueppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; LKLF Krueppel-like factor 2 Kruppel-like factor 2 (lung) Kruppel-like factor LKLF LKLFLung krueppel-like factor lung Kruppel-like zinc finger transcription factor; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor
Gene Aliases: KLF2; LKLF
UniProt ID: (Human) Q9Y5W3, (Mouse) Q60843, (Rat) Q9ET58
Entrez Gene ID: (Sheep) 101118186, (Human) 10365, (Mouse) 16598, (Rat) 306330
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.