Invitrogen
Manufacturer Code:PA544023
Catalog # PIPA544023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KLB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-klotho; betaKlotho; BKL; klotho beta; Klotho beta BKL MGC142213; klotho beta like; klotho beta, pseudogene 1; Klotho beta-like protein
Gene Aliases: AV071179; Betakl; betaKlotho; BKL; KLB; Klb-ps1; RGD1308227
UniProt ID: (Human) Q86Z14, (Mouse) Q99N32
Entrez Gene ID: (Dog) 479110, (Human) 152831, (Bovine) 514925, (Horse) 100064228, (Rabbit) 100354505, (Pig) 100525825, (Mouse) 83379, (Rat) 289625, (Guinea pig) 100734542
Molecular Function:
glucosidase
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.