Invitrogen
Manufacturer Code:PA5116394
Catalog # PIPA5116394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-29 amino acids from human KLB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-klotho; betaKlotho; betaKlotho Beta-klotho BKL KLB klotho beta klotho beta like klotho beta-like protein MGC142213; BKL; klotho beta like; Klotho beta-like protein
Gene Aliases: BKL; KLB
UniProt ID: (Human) Q86Z14
Entrez Gene ID: (Human) 152831
Molecular Function:
glucosidase
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.