Invitrogen
Manufacturer Code:PA533879
Catalog # PIPA533879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from C-terminus of human KISS1R / GPR54. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; hOT7T175; Hypogonadotropin-1; KiSS-1 receptor; KiSS-1R; KISS1R AXOR12 GPR54 HOT7T175 KISS-1R KISS1 receptor G protein-coupled receptor 54 Metastin receptor Rot7t175 G-protein coupled receptor 54 HH8 Hypogonadotropin-1 KiSS-1 receptor Kisspeptins receptor; Kisspeptins receptor; Metastin receptor
Gene Aliases: AXOR12; CPPB1; GPR54; HH8; HOT7T175; KISS-1R; KISS1R
UniProt ID: (Human) Q969F8
Entrez Gene ID: (Human) 84634
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.