Invitrogen
Manufacturer Code:PA585657
Catalog # PIPA585657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center of human Kiss1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610012C01Rik; KiSS-1 metastasis-suppressor Antibody KiSS-1 Antibody HH13 Antibody; Kiss1 metastasis-suppressor; Kiss1 variant E1a-E2b-E3; Kiss1 variant E1b-E2a-E3; Kiss1 variant E1b-E2b-E3; Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-52; Kisspeptin-54; malignant melanoma metastasis-suppressor; Metastasis-suppressor KiSS-1; Metastin; Metastin45-54; MLL septin-like fusion; prepro-kisspeptin
Gene Aliases: Eseptin; HH13; KiSS-1; KISS1; kisspeptin; metastatin; PP5098
UniProt ID: (Human) Q15726, (Rat) Q7TSB7, (Mouse) Q6Y4S4
Entrez Gene ID: (Human) 3814, (Rat) 289023, (Mouse) 280287
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.