Invitrogen
Manufacturer Code:PA572252
Catalog # PIPA572252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 115-145 amino acids from the human region of human KISS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-54; malignant melanoma metastasis-suppressor; Metastasis-suppressor KiSS-1; Metastasis-suppressor KiSS-1 Kisspeptin-1 Metastin Kisspeptin-54 Kisspeptin-14 Kisspeptin-13 Kisspeptin-10 KISS1; Metastin; prepro-kisspeptin
Gene Aliases: HH13; KiSS-1; KISS1; PP5098
UniProt ID: (Human) Q15726
Entrez Gene ID: (Human) 3814
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.