Novus Biologicals
Manufacturer Code:NBP1456720.025MG
Catalog # NB018573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide surrounding amino acid 17 of mouse KiSS-1 (NP_839991). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KiSS-1 KiSS-1 metastasis-suppressor kisspeptin Kisspeptin-1 malignant melanoma metastasis-suppressor metastasis-suppressor KiSS-1 METASTIN MGC39258; Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-54; malignant melanoma metastasis-suppressor; Metastasis-suppressor KiSS-1; Metastin; prepro-kisspeptin
Gene Aliases: HH13; KiSS-1; KISS1; PP5098
UniProt ID: (Human) Q15726
Entrez Gene ID: (Human) 3814
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.