Invitrogen
Manufacturer Code:PA572824
Catalog # PIPA572824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIRREL3 antibody for a 19 amino acid peptide near the amino terminus of human KIRREL3. The immunogen is within amino acids 20 - 70 of KIRREL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kin of IRRE-like protein 3; Kin of irregular chiasm-like protein 3; KIRRE MRD4 NEPH2 PRO4502 kin of IRRE-like protein 3 kin of irregular chiasm-like protein 3 nephrin-like 2 nephrin-like protein 2; membrane protein mKirre; mKirre; nephrin-like 2; Nephrin-like protein 2; Processed kin of IRRE-like protein 3; X kin of IRRE like 3
Gene Aliases: 1500010O20Rik; 2900036G11Rik; KIAA1867; KIRRE; KIRREL3; mKIAA1867; MRD4; NEPH2; PRO4502; SST4; UNQ5923/PRO4502/PRO19814
UniProt ID: (Human) Q8IZU9, (Mouse) Q8BR86
Entrez Gene ID: (Human) 84623, (Mouse) 67703, (Rat) 315546
Molecular Function: cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.