Invitrogen
Manufacturer Code:MA531788
Catalog # PIMA531788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6D9F6 |
Immunogen | Purified recombit fragment of human KIR3DL1 (AA: extra 22-340) expressed in Hek293 cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD158 antigen-like family member E; CD158e; HLA-BW4-specific inhibitory NK cell receptor; Killer cell immunoglobulin-like receptor 3DL1; killer cell immunoglobulin-like receptor three domains long cytoplasmic tail 2; killer cell immunoglobulin-like receptor three domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1; killer Ig receptor; KIR antigen 3DL1; KIR NKB1 NKAT3 NKB1B NKAT-3 CD158E1 KIR3DL1/S1; MHC class I NK cell receptor; Natural killer-associated transcript 3; NK-receptor; NKAT-3; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clones CL-2/CL-11; p70 NK receptor CL-2/CL-11
Gene Aliases: CD158E; CD158E1; KIR; KIR3DL1; KIR3DL1/S1; KIR3DL2; NKAT-3; NKAT3; NKB1; NKB1B
UniProt ID: (Human) P43629, (Rat) P83556
Entrez Gene ID: (Human) 3811, (Rat) 353253
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.