R&D Systems
Manufacturer Code:FAB12251P025
Catalog # FAB12251P02
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 177407 |
Immunogen | BaF3 mouse pro-B cell line transfected with human KIR3DL1 Accession # P43629 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMB11 CD158 antigen-like family member E CD158E CD158e antigen CD158e1 CD158e1/2 CD158e2 cl-11 cl-2 HLA-BW4-specific inhibitory NK cell receptor killer cell immunoglobulin-like receptor 3DL1 killer cell immunoglobulin-like receptor three domains long cytoplasmic tail1nkat3 killer Ig receptor KIR antigen 3DL1 KIR3DL1/S1 KIR3DS1 MGC119726 MGC119728 MGC126589 MGC126591 MHC class I NK cell receptor Natural killer-associated transcript 3 Nkat3 NKAT-3 NKAT3KIR NKB1 NKB1B NKB1CD158E1 NK-receptor p70 killer cell inhibitory receptor p70 natural killer cell receptor clones CL-2/CL-11 p70 NK receptor CL-2/CL-11; CD158 antigen-like family member E; CD158e; HLA-BW4-specific inhibitory NK cell receptor; Killer cell immunoglobulin-like receptor 3DL1; killer cell immunoglobulin-like receptor three domains long cytoplasmic tail 2; killer cell immunoglobulin-like receptor three domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1; killer Ig receptor; KIR antigen 3DL1; MHC class I NK cell receptor; Natural killer-associated transcript 3; NK-receptor; NKAT-3; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clones CL-2/CL-11; p70 NK receptor CL-2/CL-11
Gene Aliases: CD158E; CD158E1; KIR; KIR3DL1; KIR3DL1/S1; KIR3DL2; NKAT-3; NKAT3; NKB1; NKB1B
UniProt ID: (Human) P43629
Entrez Gene ID: (Human) 3811
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.