Invitrogen
Manufacturer Code:PA576629
Catalog # PIPA576629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human KIR3DL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMB11CD158 antigen-like family member ECD158ECD158e antigenCD158E1CD158E1/2CD158E2CL11CL2HLA-BW4-specific inhibitory NK cell receptorKI3L1&lowbarHUMANkiller cell immunoglobulin like receptorKiller cell immunoglobulin like receptor three domains short cytoplasmic tail 1Killer cell immunoglobulin like receptor three domains long cytoplasmic tail 1Killer cell immunoglobulin-like receptor 3DL1KIRKIR antigen 3DL1KIR G1Kir3dl1KIR3DS1Kirl1Kirl2Krl1MGC119726MGC119728MGC126589MGC126591MHC class I NK cell receptorNatural killer associated transcript 3Natural killer cell inhibitory receptorNatural killer-associated transcript 3NK receptorNK-associated transcript 10NK-associated transcript 3NK-associated transcript 3delIg1NKAT-3NKAT10NKAT3NKB1NKB1Bp70 killer cell inhibitory receptorp70 natural killer cell receptor clones CL 2/CL 11p70 natural killer cell receptor clones CL-2/CL-11p70 NK receptor CL-2/CL-11; CD158 antigen-like family member E; CD158e; HLA-BW4-specific inhibitory NK cell receptor; Killer cell immunoglobulin-like receptor 3DL1; killer cell immunoglobulin-like receptor Kir3dl1; killer cell immunoglobulin-like receptor three domains long cytoplasmic tail 2; killer cell immunoglobulin-like receptor three domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1; killer Ig receptor; killer immunoglobulin receptor-like protein 1; KIR antigen 3DL1; KIR-like receptor 1; MHC class I NK cell receptor; Natural killer-associated transcript 3; NK-receptor; NKAT-3; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clones CL-2/CL-11; p70 NK receptor CL-2/CL-11
Gene Aliases: CD158E; CD158E1; KIR; KIR3DL1; KIR3DL1/S1; KIR3DL2; Kirl1; Kirl2; Krl1; NKAT-3; NKAT3; NKB1; NKB1B
UniProt ID: (Human) P43629
Entrez Gene ID: (Human) 3811, (Mouse) 245616
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.