R&D Systems
Manufacturer Code:FAB8920N100UG
Catalog # FAB8920N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1165A |
Immunogen | NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g His22-His245 Accession # Q14953 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rabbit MonoclonalAntibody
For Research Use Only
Protein Aliases: CD158 antigen-like family member G; CD158g; Killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor 2DS5 CD158g killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 5 NKAT9; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; MHC class I NK cell receptor; natural killer cell inhibitory receptor; Natural killer-associated transcript 9; NKAT-9
Gene Aliases: CD158G; KIR2DS5; NKAT9
UniProt ID: (Human) Q14953
Entrez Gene ID: (Human) 3810
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.