Invitrogen
Manufacturer Code:MA182651
Catalog # PIMA182651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NKVFS1 |
Immunogen | Proprietary Immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD158 antigen-like family member A; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158a; CD158h; CD158i; CD158j; Killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; Killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer Ig receptor; killer inhibitory receptor 2-2-1; killer inhibitory receptor 4-1-2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR antigen 2DS4; KIR2DL KIR2DS1 KIR2DS2 p50.3 KIR2DS4 CD158a CD158b CD158 XXbac-BCX195L8.1 KIR-K64 KIR221 NKAT NKAT1 p58.1 CD158 antigen-like family member A MHC class I NK cell receptor NKAT-1 killer Ig receptor killer cell immunoglobulin-like receptor 2DL1 CD158B2 KIRCL23 NKAT2 CD158B1 NKAT6 CD158J NKAT5 CD158I KKA3 NKAT8 CD158H; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; natural killer associated transcript 5; natural killer cell inhibitory receptor; Natural killer-associated transcript 1; Natural killer-associated transcript 5; Natural killer-associated transcript 8; NK receptor 183 ActI; NKAT-1; NKAT-5; NKAT-8; p50 killer cell activating receptor KAR-K1d; p58 killer cell inhibitory receptor KIR-K64; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; P58 natural killer cell receptor clones CL-39/CL-17; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-39/CL-17; p58 NK receptor CL-42/47.11; p58 NK receptor CL-49; p58.1 MHC class-I-specific NK receptor
Gene Aliases: 183ActI; CD158A; CD158b; CD158H; CD158I; CD158J; cl-49; KIR-2DS2; KIR-2DS4; KIR-K64; KIR1D; KIR221; KIR2DL1; KIR2DP1DL1; KIR2DS1; KIR2DS2; KIR2DS4; KIR412; KKA3; NKAT; NKAT-1; NKAT-5; NKAT-8; NKAT1; NKAT5; NKAT8; p50.1; p58.1
UniProt ID: (Human) P43626, (Human) Q14954, (Human) P43631, (Human) P43632
Entrez Gene ID: (Human) 3802, (Human) 3806, (Human) 100132285, (Human) 3809
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.