Invitrogen
Manufacturer Code:PA5116207
Catalog # PIPA5116207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 151-180 amino acids from the Central region of human KIR2DS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 183ActI CD158 CD158 antigen-like family member A CD158 antigen-like family member G CD158 antigen-like family member H CD158 antigen-like family member I CD158 antigen-like family member J CD158A CD158b CD158B1 CD158G CD158h CD158i CD158J cl-49 killer cell immunoglobulin like receptor two Ig domains and long cytoplasmic tail 1 killer cell immunoglobulin like receptor two Ig domains and short cytoplasmic tail 1 killer cell immunoglobulin like receptor two Ig domains and short cytoplasmic tail 2 killer cell immunoglobulin like receptor two Ig domains and short cytoplasmic tail 4 killer cell immunoglobulin like receptor two Ig domains and short cytoplasmic tail 5 killer cell immunoglobulin-like receptor 2DL1 Killer cell immunoglobulin-like receptor 2DS1 killer cell immunoglobulin-like receptor 2DS2 Killer cell immunoglobulin-like receptor 2DS4 killer cell immunoglobulin-like receptor 2DS5 killer cell immunoglobulin-like receptor KIR2DS2 killer cell immunoglobulin-like receptor KIRDS1 killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1 killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 1 killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1 killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2 killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 4 killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 5 killer Ig receptor killer inhibitory receptor 2-2-1 killer inhibitory receptor 4-1-2 killer-cell Ig-like receptor killer-cell immunoglobulin-like receptor killer-cell immunoglobulin-like receptor two domains short tail 2 protein KIR antigen 2DS4 KIR1D KIR221 KIR2DL1 KIR2DP1DL1 KIR2DS1 KIR2DS2 KIR-2DS2 KIR2DS4 KIR-2DS4 KIR2DS5 KIR412 KIR-K64 KKA3 MHC class I NK cell receptor MHC class I NK cell receptor Eb6 ActI natural killer associated transcript 5 natural killer cell inhibitory receptor natural killer cell receptor natural killer-associated transcript 1 Natural killer-associated transcript 5 Natural killer-associated transcript 8 natural killer-associated transcript 9 NK receptor 183 ActI NKAT NKAT1 NKAT-1 NKAT5 NKAT-5 NKAT6 NKAT8 NKAT-8 NKAT9 NKAT-9 p50 killer cell activating receptor KAR-K1d p50.1 p50.3 p58 killer cell inhibitory receptor KIR-K64 p58 killer cell inhibitory receptor KIR-K7a p58 natural killer cell receptor clone CL-49 P58 natural killer cell receptor clones CL-39/CL-17 p58 natural killer cell receptor clones CL-42/47.11 p58 NK cell inhibitory receptor NKR-K6 p58 NK receptor CL-39/CL-17 p58 NK receptor CL-42/47.11 p58 NK receptor CL-49 p58.1 p58.1 MHC class-I-specific NK receptor XXbac-BCX195L8.1; CD158 antigen-like family member A; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158a; CD158g; CD158h; CD158i; CD158j; Killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; Killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; Killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig receptor; killer inhibitory receptor 2-2-1; killer inhibitory receptor 4-1-2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR antigen 2DS4; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; natural killer associated transcript 5; natural killer cell inhibitory receptor; Natural killer-associated transcript 1; Natural killer-associated transcript 5; Natural killer-associated transcript 8; Natural killer-associated transcript 9; NK receptor 183 ActI; NKAT-1; NKAT-5; NKAT-8; NKAT-9; p50 killer cell activating receptor KAR-K1d; p58 killer cell inhibitory receptor KIR-K64; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; P58 natural killer cell receptor clones CL-39/CL-17; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-39/CL-17; p58 NK receptor CL-42/47.11; p58 NK receptor CL-49; p58.1 MHC class-I-specific NK receptor
Gene Aliases: 183ActI; CD158A; CD158b; CD158G; CD158H; CD158I; CD158J; cl-49; KIR-2DS2; KIR-2DS4; KIR-K64; KIR1D; KIR221; KIR2DL1; KIR2DP1DL1; KIR2DS1; KIR2DS2; KIR2DS4; KIR2DS5; KIR412; KKA3; NKAT; NKAT-1; NKAT-5; NKAT-8; NKAT1; NKAT5; NKAT8; NKAT9; p50.1; p58.1
UniProt ID: (Human) P43626, (Human) Q14954, (Human) P43631, (Human) P43632, (Human) Q14953
Entrez Gene ID: (Human) 3802, (Human) 3806, (Human) 100132285, (Human) 3809, (Human) 3810
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.