Novus Biologicals
Manufacturer Code:NB10063267
Catalog # NB10063267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NKVFS1 |
Immunogen | Made to Human KIR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD158 antigen-like family member A; CD158 antigen-like family member A CD158a antigen CD158Ankat1 cl-42 killer cell immunoglobulin-like receptor 2DL1 killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 1 killer Ig receptor killer inhibitory receptor 2-2-1 KIR221 MHC class I NK cell receptor Natural killer-associated transcript 1 NKAT NKAT-1 NKAT1KIR-K64 p58 killer cell inhibitory receptor KIR-K64 p58 natural killer cell receptor clones CL-42/47.11 p58 NK cell inhibitory receptor NKR-K647.11 p58 NK receptor CL-42/47.11 p58.1 p58.1 MHC class-I-specific NK receptor; CD158a; Killer cell immunoglobulin-like receptor 2DL1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer Ig receptor; killer inhibitory receptor 2-2-1; MHC class I NK cell receptor; Natural killer-associated transcript 1; NKAT-1; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1 MHC class-I-specific NK receptor
Gene Aliases: CD158A; KIR-K64; KIR221; KIR2DL1; KIR2DS1; NKAT; NKAT-1; NKAT1; p58.1
UniProt ID: (Human) P43626
Entrez Gene ID: (Human) 3802
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.