Proteintech
Manufacturer Code:602661IG150UL
Catalog # 50-173-6363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7C9F2 |
Immunogen | KIFAP3 Fusion Protein Ag20809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: KAP 3 KAP3 KIF3AP KIFAP3 kinesin associated protein 3 SMAP Smg GDS Smg GDS associated protein; KAP-3; Kinesin-associated protein 3; small G protein GDP dissociation stimulator; Smg GDS-associated protein
Gene Aliases: dJ190I16.1; FLA3; KAP-1; KAP-3; KAP3; KIF3AP; KIFAP3; SMAP; Smg-GDS
UniProt ID: (Human) Q92845, (Mouse) P70188
Entrez Gene ID: (Human) 22920, (Rat) 289168, (Mouse) 16579
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.