Proteintech
Manufacturer Code:216321AP150UL
Catalog # 50-173-3522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIF5B Fusion Protein Ag16250 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Conventional kinesin heavy chain; epididymis secretory protein Li 61; KIF5B Kinesin 1 heavy chain kinesin family member 5B KINH KNS KNS1 Ubiquitous kinesin heavy chain UKHC; kif5b {ECO:0000312|EMBL:ABC25059.1}; kinesin 1 (110-120kD); kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; Kinesin-1 heavy chain; Ubiquitous kinesin heavy chain; UKHC
Gene Aliases: AL022807; HEL-S-61; Khc; Khcs; KIF5B; KINH; KNS; KNS1; UKHC
UniProt ID: (Human) P33176, (Mouse) Q61768, (Rat) Q2PQA9
Entrez Gene ID: (Human) 3799, (Mouse) 16573, (Rat) 117550
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.