Invitrogen
Manufacturer Code:PA5115469
Catalog # PIPA5115469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KIF4A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323435 Chromokinesin chromokinesin-A Chromosome-associated kinesin KIF4 Chromosome-associated kinesin KIF4A D330050K22Rik FLJ12530 FLJ12655 FLJ14204 FLJ20631 HSA271784 KIF4 KIF4A KIF4G1 KIF4-G1 kinesin family member 4 kinesin family member 4A kinesin heavy chain member 4 Kns4 MRX100 N-5 kinesin; Chromokinesin-A; Chromosome-associated kinesin KIF4A
Gene Aliases: KIF4; KIF4A; KIF4G1; MRX100
UniProt ID: (Human) O95239
Entrez Gene ID: (Human) 24137
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.