Proteintech
Manufacturer Code:143441AP150UL
Catalog # 50-172-9381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIF4A Fusion Protein Ag5682 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromokinesin; Chromokinesin A HSA271784 KIF4 KIF4 G1 KIF4A kinesin family member 4A; Chromokinesin-A; Chromosome-associated kinesin KIF4; Chromosome-associated kinesin KIF4A; kinesin family member 4; kinesin family member 4A; kinesin heavy chain member 4; N-5 kinesin
Gene Aliases: AI323435; D330050K22Rik; KIF4; KIF4A; KIF4G1; Kns4; MRX100
UniProt ID: (Human) O95239, (Mouse) P33174
Entrez Gene ID: (Human) 24137, (Mouse) 16571, (Rat) 84393
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.