Invitrogen
Manufacturer Code:PA552386
Catalog # PIPA552386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human KIF3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HH0048; Kinesin-like protein KIF3B; Kinesin-like protein KIF3B, N-terminally processed; Microtubule plus end-directed kinesin motor 3B
Gene Aliases: AI854312; AW549267; FLA8; HH0048; KIAA0359; KIF3B; KLP-11; mKIAA0359
UniProt ID: (Human) O15066, (Mouse) Q61771
Entrez Gene ID: (Human) 9371, (Mouse) 16569, (Rat) 296284
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.