Novus Biologicals
Manufacturer Code:NBP305143100UL
Catalog # NB094624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 663-747 of human KIF3B (NP_004789.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HH0048; HH0048 KIAA0359Microtubule plus end-directed kinesin motor 3B kinesin family member 3B kinesin-like protein KIF3B; Kinesin-like protein KIF3B; Kinesin-like protein KIF3B, N-terminally processed; Microtubule plus end-directed kinesin motor 3B
Gene Aliases: FLA8; HH0048; KIAA0359; KIF3B; KLP-11
UniProt ID: (Human) O15066
Entrez Gene ID: (Human) 9371
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.