OriGene
Manufacturer Code:TA503320
Catalog # 50-167-4246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C5 |
Immunogen | Full length human recombit protein of human KIF2C produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CT139 KNSL6 MCAK; kinesin-like 6; Kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; MCAK; Mitotic centromere-associated kinesin; testis tissue sperm-binding protein Li 68n
Gene Aliases: 4930402F02Rik; CT139; ESTM5; KIF2C; KNSL6; KRP2; MCAK; X83316
UniProt ID: (Human) Q99661, (Mouse) Q922S8
Entrez Gene ID: (Human) 11004, (Mouse) 73804, (Rat) 171529
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.