Invitrogen
Manufacturer Code:PA568623
Catalog # PIPA568623
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human KIF23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHO1 KNSL5 MKLP-1 MKLP1 kinesin-like 5 (mitotic kinesin-like protein 1) kinesin-like protein 5 kinesin-like protein KIF23 mitotic kinesin-like 1 mitotic kinesin-like protein 1; kinesin-like 5; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Kinesin-like protein KIF23-like protein; Mitotic kinesin-like protein 1
Gene Aliases: 3110001D19Rik; C87313; cb738; CHO1; KIF23; KNSL5; MKLP-1; MKLP1; wu:fi30e02; wu:fi39f08; zMKLP1
UniProt ID: (Human) Q02241, (Mouse) E9Q5G3
Entrez Gene ID: (Bovine) 515748, (Horse) 100064697, (Pig) 100522116, (Human) 9493, (Dog) 478355, (Rabbit) 100343765, (Rat) 315740, (Mouse) 71819, (Guinea pig) 100714156, (Zebrafish) 30627
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.