Invitrogen
Manufacturer Code:PA567017
Catalog # PIPA567017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human KIF23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHO1 KNSL5 MKLP-1 MKLP1 kinesin-like 5 (mitotic kinesin-like protein 1) kinesin-like protein 5 kinesin-like protein KIF23 mitotic kinesin-like 1 mitotic kinesin-like protein 1; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Mitotic kinesin-like protein 1
Gene Aliases: CHO1; KIF23; KNSL5; MKLP-1; MKLP1
UniProt ID: (Human) Q02241
Entrez Gene ID: (Human) 9493
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.