Invitrogen
Manufacturer Code:PA5100863
Catalog # PIPA5100863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KIF23 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3110001D19Rik C87313 CHO1 KIF23 kinesin family member 23 kinesin-like 5 (mitotic kinesin-like protein 1) Kinesin-like protein 5 Kinesin-like protein KIF23 KNSL5 Mitotic kinesin-like protein 1 MKLP1 MKLP-1; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Mitotic kinesin-like protein 1
Gene Aliases: 3110001D19Rik; C87313; CHO1; KIF23; KNSL5; MKLP-1; MKLP1
UniProt ID: (Human) Q02241, (Mouse) E9Q5G3
Entrez Gene ID: (Human) 9493, (Mouse) 71819
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.