Proteintech
Manufacturer Code:195511AP150UL
Catalog # 50-173-2451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIF19 KIF19A kinesin family member 19 Kinesin like protein KIF19; KIF19A-like protein; Kinesin-like protein KIF19; N-8 kinesin; oocyte-G1
Gene Aliases: KIF19; KIF19A
UniProt ID: (Human) Q52M87, (Mouse) Q99PT9
Entrez Gene ID: (Human) 124602, (Mouse) 286942
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.