Invitrogen
Manufacturer Code:PA530874
Catalog # PIPA530874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 725 and 1015 of Human KIF13A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of mouse KIF13A mannose-6-phosphate receptor transporter; Kinesin-like protein KIF13A; Kinesin-like protein RBKIN; RBKIN bA500C11.2 homolog of mouse KIF13A mannose-6-phosphate receptor transporter kinesin-like protein KIF13A kinesin-like protein RBKIN
Gene Aliases: bA500C11.2; KIF13A; RBKIN
UniProt ID: (Human) Q9H1H9
Entrez Gene ID: (Human) 63971, (Rat) 308173
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.