Proteintech
Manufacturer Code:272101AP150UL
Catalog # 50-173-5603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIF13A Fusion Protein Ag24356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of mouse KIF13A mannose-6-phosphate receptor transporter; KIF13A kinesin family member 13A Kinesin like protein KIF13A Kinesin like protein RBKIN RBKIN; Kinesin-like protein KIF13A; Kinesin-like protein RBKIN
Gene Aliases: bA500C11.2; KIF13A; RBKIN
UniProt ID: (Human) Q9H1H9
Entrez Gene ID: (Human) 63971
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.