Novus Biologicals
Manufacturer Code:NBP190961
Catalog # NBP190961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LRHQPILMGAHLAVIDALMAAFAFEWTKTLPGPLALTSLEHKLLFWVDTTVRRLQEKTEQEAAQRASPAAPADGAAPAQPSIRYRKDR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calmodulin regulated spectrin-associated protein family member 3 calmodulin-regulated spectrin-associated protein 3 KIAA1543 NEZHA protein Nezha; calmodulin regulated spectrin-associated protein family, member 3; Calmodulin-regulated spectrin-associated protein 3; Protein Nezha; protein phosphatase 1, regulatory subunit 80
Gene Aliases: CAMSAP3; KIAA1543; NEZHA; PPP1R80
UniProt ID: (Human) Q9P1Y5
Entrez Gene ID: (Human) 57662
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.